Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFkB p100/p52, Rabbit anti-Human, Clone: SAIC-26C-15, Novus Biologicals™
SDP

Catalog No. NB016834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016834 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016834 Supplier Novus Biologicals Supplier No. NBP2812720.2MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NFkB p100/p52 Monoclonal specifically detects NFkB p100/p52 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Functional.

Specifications

Antigen NFkB p100/p52
Applications Functional Assay
Classification Monoclonal
Clone SAIC-26C-15
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Functional
Formulation PBS
Gene Alias DNA-binding factor KBF2, H2TF1, Lymphocyte translocation chromosome 10 protein, Lyt10, LYT10LYT-10, nuclear factor NF-kappa-B p100 subunit, nuclear factor of kappa light chain gene enhancer in B-cells 2, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100), Oncogene Lyt-10, p52
Gene Symbols NFKB2
Host Species Rabbit
Immunogen Peptide IEVDLVTHSDPPR derived from the transcription factor NFKB2 (Nuclear factor NF-kappa-B p100 subunit) conjugated to KLH.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4791
Test Specificity Recognises human NFKB2.
Target Species Human
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.