Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFkB p105/p50 Antibody (2J10D7), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10056583R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056583R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056583R Supplier Novus Biologicals Supplier No. NB10056583R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NFkB p105/p50 Monoclonal specifically detects NFkB p105/p50 in Human, Rabbit, Rat samples. It is validated for Flow Cytometry, Gene Knock-Out, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen NFkB p105/p50
Applications Flow Cytometry, Gene Knock-Out, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2J10D7
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated
Gene Accession No. P25942
Gene Alias DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50
Gene Symbols NFKB1
Host Species Mouse
Immunogen A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4790
Test Specificity Cross reacts with Human. Predicted to react with Mouse and Rat which are 86% homologous with the immunogen.
Target Species Human, Rabbit, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.