Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NFkB p50 (NLS) Inhibitor Peptide Set
SDP

Applications: Functional (Inhibition)

$706.00 - $1244.00

Specifications

Host Species Human, Mouse, Rat, Bovine, Canine, Chicken, Xenopus
Components NF-kB p50 (NLS) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKVQRKRQKLM (p50 NLS sequence: VQRKRQKLM). Molecular weight: 3529. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
For Use With (Application) Functional (Inhibition)
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Product Type NFkB p105/p50 Inhibitor Peptide Set
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP229323
SDP
View Documents
Novus Biologicals™
NBP229323
2 mg
Each for $706.00
Only null left
Add to Cart
 
NBP2293235
SDP
View Documents
Novus Biologicals™
NBP2293235MG
5 mg
Each for $1,244.00
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Human, Mouse, Rat, Bovine, Canine, Chicken, Xenopus
Functional (Inhibition)
NFkB p105/p50 Inhibitor Peptide Set
NFkB p105/p50
NF-kB p50 (NLS) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKVQRKRQKLM (p50 NLS sequence: VQRKRQKLM). Molecular weight: 3529. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
3529
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.