Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NFPS

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, non-transportable inhibitor of GlyT1 which displays no activity at GABA or glutamate receptors (IC50 values are 2.8, 9.8 and 56000 nM for hGlyT1, rGlyT1 and rGlyT2 respectively). Enhances the amplitude of the NMDA component of glutamatergic EPSCs.
R-enantiomer also available.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 393.45 |
Purity | >98% |
Molecular Weight (g/mol) | 393.458 |
Chemical Name or Material | NFPS |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction