Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFS1 Antibody (OTI2H5), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP272978A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272978A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272978A70 Supplier Novus Biologicals Supplier No. NBP272978AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NFS1 Monoclonal specifically detects NFS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NFS1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2H5
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cysteine desulfurase, mitochondrial, EC 2.8.1.7, NFS1 nitrogen fixation 1 homolog (S. cerevisiae), NifS, NIFShomolog), nitrogen-fixing bacteria S-like protein
Gene Symbols NFS1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-299 of human NFS1 (NP_066923) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9054.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.