Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFX1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291347 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291347 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291347 Supplier Novus Biologicals Supplier No. NBP149932APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFX1 Polyclonal antibody specifically detects NFX1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen NFX1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias DKFZp779G2416, EC 6.3.2.-, MGC20369, NFX2transcriptional repressor NF-X1, Nuclear transcription factor, X box-binding protein 1, nuclear transcription factor, X-box binding 1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 250 and 300 of human Nuclear Transcription Factor, X-box Binding 1 using the numbering given in entry NP_002495.2 (GeneID 4799).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 4799
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.