Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFX1 Antibody, Novus Biologicals™
SDP

Catalog No. NB394109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB394109 25 μL
NBP257148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB394109 Supplier Novus Biologicals Supplier No. NBP25714825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFX1 Polyclonal specifically detects NFX1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NFX1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias DKFZp779G2416, EC 6.3.2.-, MGC20369, NFX2transcriptional repressor NF-X1, Nuclear transcription factor, X box-binding protein 1, nuclear transcription factor, X-box binding 1
Gene Symbols NFX1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VFHPDSSEASSRKGVLDGYGARRNEQRRYPQKRPPWEVEGARPRPGRNPPKQEGHRHTNAGHRNNMGPIPKDDLNERPAKSTCDSENLAVINKSSR
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 4799
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.