Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2/MCSP Antibody (LHM 2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB1002688J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002688J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002688J Supplier Novus Biologicals Supplier No. NB1002688APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NG2/MCSP Monoclonal specifically detects NG2/MCSP in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Cell Surface), Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen NG2/MCSP
Applications Flow Cytometry, Flow Cytometry (Cell Surface), Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LHM 2
Conjugate APC
Dilution Flow Cytometry, Knockout Validated
Gene Accession No. Q6UVK1
Gene Alias chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), EC 3.6.3, HMW-MAA, MCSPChondroitin sulfate proteoglycan NG2, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8
Gene Symbols CSPG4
Host Species Mouse
Immunogen A 375P cells crude extract
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1464
Test Specificity Human. Other species have not been tested.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.