Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2/MCSP Antibody (LHM 2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013946 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013946 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013946 Supplier Novus Biologicals Supplier No. NBP280873
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NG2/MCSP Monoclonal specifically detects NG2/MCSP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Knockout Validated.

Specifications

Antigen NG2/MCSP
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LHM 2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:1000, Flow Cytometry, Immunohistochemistry : 1:200, Immunocytochemistry/Immunofluorescence : 1:100, Immunoprecipitation : 1:10 - 1:100, Immunohistochemistry-Paraffin : 1:200, Immunohistochemistry-Frozen : 1:200, Flow (Cell Surface) 1 - 2.5 μg/mL, Knockout Validated
Formulation Tris-Glycine, 0.15 M NaCl
Gene Alias chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), EC 3.6.3, HMW-MAA, MCSPChondroitin sulfate proteoglycan NG2, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8
Gene Symbols CSPG4
Host Species Mouse
Immunogen A 375P cells crude extract
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1464
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.