Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NGB 2904
GREENER_CHOICESDP

Catalog No. 263510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-351-0 10 mg
26-355-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-351-0 Supplier Tocris Bioscience™ Supplier No. 2635/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective D3 antagonist

NGB 2904 is a potent and selective dopamine D3 receptor antagonist (Ki values are 1.4, 217, 223, 642, > 5000, > 10000 and > 10000 nM for D3, D2, 5-HT2, α1, D4, D1 and D5 receptors respectively). Potently antagonizes quinpirole -stimulated mitogenesis (IC50 = 6.8 nM). Attenuates cocaine's rewarding effects and inhibits relapse to drug-seeking behavior.

Chemical Identifiers

CAS 189061-11-8
Molecular Formula C28H30Cl3N3O

Specifications

Quantity 10 mg
Formula Weight 530.92
Purity >98%
Molecular Weight (g/mol) 530.918
Chemical Name or Material NGB 2904
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.