Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGDN Antibody (OTI1H4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB272979A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272979A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272979A48 Supplier Novus Biologicals Supplier No. NBP272979AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NGDN Monoclonal specifically detects NGDN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NGDN
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1H4
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C14orf120, DKFZp564O092, EIF4E-binding protein, eukaryotic initiation factor 4E and CPEB binding protein, LCP5, lpd-2, neuroguidin, neuroguidin, EIF4E binding protein, NGDchromosome 14 open reading frame 120
Gene Symbols NGDN
Host Species Mouse
Immunogen Full length recombinant protein of human NGDN (NP_001036100) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 25983.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.