Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFI-B alpha/Nur77/NR4A1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 21 publication

Supplier:  Novus Biologicals NB10056745

 View more versions of this product

Catalog No. NB10056745


Only null left
Add to Cart

Description

Description

NGFI-B alpha/Nur77/NR4A1 Polyclonal specifically detects NGFI-B alpha/Nur77/NR4A1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

NGFI-B alpha/Nur77/NR4A1
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor
Rabbit
Immunogen affinity purified
RUO
Primary
Human and Mouse. Not yet tested in other species.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
1.0 mg/mL
Western Blot 1:500-1:1000, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Lee et al (2009)), Immunohistochemistry 1:200. Use reported in scientific literature (PMID 31115481), Immunocytochemistry/Immunofluorescence 1:10-1:2000. Use reported in scientific literature (Chintharlapalli et al (2005)), Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Ferlini et al (2009)), Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000
P22736
NR4A1
This antibody was developed against a synthetic peptide corresponding to aa 251-266 of human Nak1 (NP_775180).
0.1 mg
Apoptosis
3164
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only