Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFI-B alpha/Nur77/NR4A1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB339458 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339458 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339458 Supplier Novus Biologicals Supplier No. NB10056745MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NGFI-B alpha/Nur77/NR4A1 Polyclonal antibody specifically detects NGFI-B alpha/Nur77/NR4A1 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)

Specifications

Antigen NGFI-B alpha/Nur77/NR4A1
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the human NGFI-B alpha protein (between amino acids 200-300) [UniProt P22736].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 3164
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.