Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGFI-B alpha/Nur77/NR4A1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | NGFI-B alpha/Nur77/NR4A1 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NGFI-B alpha/Nur77/NR4A1 Polyclonal antibody specifically detects NGFI-B alpha/Nur77/NR4A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NGFI-B alpha/Nur77/NR4A1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS (pH 7.2) and 40% Glycerol | |
3164 | |
IgG | |
Protein A purified |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor | |
This antibody was developed against a recombinant protein corresponding to amino acids: PANLLTSLVRAHLDSGPSTAKLDYSKFQELVLPHFGKEDAGDVQQFYDLLS | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title