Learn More
NGFR (extracellular) Monoclonal Antibody (192-IgG), Invitrogen™
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 100 μL of deionized water.
Specifications
Specifications
| Antigen | NGFR (extracellular) |
| Applications | Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 192-IgG |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | Ngfr |
| Gene Accession No. | P07174 |
| Gene Alias | NGFR, CD271, TNFR superfamily, member 16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.