Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFR/TNFRSF16/p75NTR Antibody (NGFR/2550R), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB379817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379817 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379817 Supplier Novus Biologicals Supplier No. NBP308566AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NGFR/TNFRSF16/p75NTR Monoclonal antibody specifically detects NGFR/TNFRSF16/p75NTR in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen NGFR/TNFRSF16/p75NTR
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NGFR/2550R
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16
Host Species Rabbit
Immunogen Recombinant human NGFR/TNFRSF16/p75NTR protein fragment (around aa 281-421) (exact sequence is proprietary) (Uniprot: P08138)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4804
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.