Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFR/TNFRSF16/p75NTR Antibody (NGFR5 + NTR/912), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP247967A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP247967A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP247967A70 Supplier Novus Biologicals Supplier No. NBP247967AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NGFR/TNFRSF16/p75NTR Monoclonal specifically detects NGFR/TNFRSF16/p75NTR in Human, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen NGFR/TNFRSF16/p75NTR
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone NGFR5 + NTR/912
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16
Gene Symbols NGFR
Host Species Mouse
Immunogen NGFR from A875 melanoma cells (NGFR5); Recombinant human p75 NGFR protein (NTR/912)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4804
Test Specificity It recognizes a glycoprotein of 75kDa, identified as low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR). NGFR is expressed in various neural crest cells and their tumors such as melanocytes, melanomas, neuroblastomas, pheochromocytomas and neurofibromas. Reportedly, anti-NGFR is a reliable marker for desmoplastic and neurotropic melanomas. NGFR is expressed in mature non-neural cells such as perivascular cells, dental pulp cells, lymphoidal follicular dendritic cells, basal epithelium of oral mucosa and hair follicles, prostate basal cells, and myoepithelial cells. Anti-NGFR stains the myoepithelial cells of breast ducts and intra-lobular fibroblasts of breast ducts.
Target Species Human, Primate, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.