Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFRAP1/BEX3/NADE Antibody, Novus Biologicals™
SDP

Catalog No. NBP131652 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131652 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131652 Supplier Novus Biologicals Supplier No. NBP131652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NGFRAP1/BEX3/NADE Polyclonal specifically detects NGFRAP1/BEX3/NADE in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NGFRAP1/BEX3/NADE
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.38mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
Gene Alias BEX3p75NTR-associated cell death executor, Brain-expressed X-linked protein 3, DXS6984EX-linked 3, HGR74, nerve growth factor receptor (TNFRSF16) associated protein 1, Nerve growth factor receptor-associated protein 1, Ovarian granulosa cell 13.0 kDa protein HGR74, ovarian granulosa cell protein (13kD), protein BEX3
Gene Symbols BEX3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NADE. The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 27018
Test Specificity Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.