Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGL-1/LRRC4C Antibody, Novus Biologicals™
SDP

Catalog No. NBP219515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219515 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219515 Supplier Novus Biologicals Supplier No. NBP219515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NGL-1/LRRC4C Polyclonal specifically detects NGL-1/LRRC4C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NGL-1/LRRC4C
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.78mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9HCJ2
Gene Alias KIAA1580NGL-1NGL1Netrin-G1 ligand, leucine rich repeat containing 4C, leucine-rich repeat-containing protein 4C
Gene Symbols LRRC4C
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human Netrin G1 ligand. The exact sequence is proprietary.
Molecular Weight of Antigen 72 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57689
Test Specificity Human. Species reactivity based on sequence identity: Mouse (99%), Rat (99%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.