Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NGL-3/LRRC4B/LRCH4B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP231736
Description
NGL-3/LRRC4B/LRCH4B Polyclonal specifically detects NGL-3/LRRC4B/LRCH4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NGL-3/LRRC4B/LRCH4B | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9NT99 | |
LRRC4B | |
This antibody was developed against a recombinant protein corresponding to amino acids: GTEKEPPGPTTDGVWGGGRPGDAAGPASSSTTAPAPRSSRPTEKAFTVPITDVTENALKDLDD | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DKFZp761A179, HSM, Leucine Rich Repeat Containing 4B, Leucine-Rich Repeat-Containing Protein 4B, Leucine-Rich Repeats And Immunoglobulin-Like Domains 4, LRIG4, LRRC4B, Netrin-G3 Ligand, NGL-3 | |
Rabbit | |
Affinity Purified | |
RUO | |
94030 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction