Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NgR2/NgRH1/RTN4RL2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343774 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343774 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343774 Supplier Novus Biologicals Supplier No. NBP241200MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NgR2/NgRH1/RTN4RL2 Polyclonal antibody specifically detects NgR2/NgRH1/RTN4RL2 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen NgR2/NgRH1/RTN4RL2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias NgR2, NGRH1, reticulon 4 receptor-like 2
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human RTN4RL2. The immunogen is located within amino acids 330 - 380 of RTN4RL2. Amino Acid Squence: LYRDLPAEDSRGRQGGDA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 349667
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.