Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NgR2/NgRH1/RTN4RL2 Antibody (P14), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP22676A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22676A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22676A94 Supplier Novus Biologicals Supplier No. NBP252676AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NgR2/NgRH1/RTN4RL2 Monoclonal specifically detects NgR2/NgRH1/RTN4RL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen NgR2/NgRH1/RTN4RL2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone P14
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias NgR2, NGRH1, reticulon 4 receptor-like 2
Gene Symbols RTN4RL2
Host Species Rabbit
Immunogen (His)6-tagged fragment of rat NgR2 (amino acids 279-420).
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 349667
Test Specificity The antibody binds to human NgR2 with a Kd of 2.3 nM and to rat NgR2 with a Kd of 0.48 nM (bivalent binding).
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.