Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGX6 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB051510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB051510 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB051510 Supplier Novus Biologicals Supplier No. NBP298040FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NGX6 Polyclonal specifically detects NGX6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NGX6
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C9orf127, chromosome 9 open reading frame 127, NAG-5, nasopharyngeal carcinoma expressed 6, nasopharyngeal carcinoma related protein, Nasopharyngeal carcinoma-associated gene 6 protein, NGX6MGC120460, Protein NAG-5, Protein NGX6, RP11-112J3.10, transmembrane protein 8B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NGX6 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 51754
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.