Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NH 125
GREENER_CHOICE

Catalog No. 343910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-391-0 10 mg
34-395-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 34-391-0 Supplier Tocris Bioscience™ Supplier No. 3439/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

CaM kinase III (eEF-2 kinase) inhibitor

Eukaryotic elongation factor 2 (eEF-2) kinase (CaMK III) inhibitor (IC50 = 60 nM) that displays 125-fold, > 1300-fold and > 1500-fold selectivity over PKC, PKA and CaMK II respectively. Decreases the viability of a variety of cancer cell lines (IC50 values are 0.7 - 4.8 μM) and blocks G1/S cell cycle progression. Shown to induce eEF-2 phosphorylation in cancer cells. Also an effective antibacterial agent; inhibits histidine protein kinase in vitro (IC50 = 6.6 μM) and in vivo.

Chemical Identifiers

CAS 278603-08-0
Molecular Formula C27H45IN2

Specifications

Quantity 10 mg
Formula Weight 524.56
Purity >98%
Molecular Weight (g/mol) 524.575
Chemical Name or Material NH 125
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.