Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NH 125
GREENER_CHOICE

Catalog No. p-4817088
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CaM kinase III (eEF-2 kinase) inhibitor

Eukaryotic elongation factor 2 (eEF-2) kinase (CaMK III) inhibitor (IC50 = 60 nM) that displays 125-fold, > 1300-fold and > 1500-fold selectivity over PKC, PKA and CaMK II respectively. Decreases the viability of a variety of cancer cell lines (IC50 values are 0.7 - 4.8 μM) and blocks G1/S cell cycle progression. Shown to induce eEF-2 phosphorylation in cancer cells. Also an effective antibacterial agent; inhibits histidine protein kinase in vitro (IC50 = 6.6 μM) and in vivo.

Chemical Identifiers

CAS 278603-08-0
Molecular Formula C27H45IN2

Specifications

Quantity 10 mg
Formula Weight 524.56
Purity >98%
Molecular Weight (g/mol) 524.575
Chemical Name or Material NH 125
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.