Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHA2/SLC9B2/NHEDC2 Antibody (896151), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346106 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346106 Supplier Novus Biologicals Supplier No. FAB8404MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NHA2/SLC9B2/NHEDC2 Monoclonal specifically detects NHA2/SLC9B2/NHEDC2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen NHA2/SLC9B2/NHEDC2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 896151
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ23984, Mitochondrial Na(+)/H(+) exchanger NHA2, mitochondrial sodium/hydrogen exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, Na+/H+ exchanger domain containing 2, NHA2NHE10, NHE domain-containing protein 2, Sodium/hydrogen exchanger-like domain-containing protein 2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NHA2/SLC9B2, Met1-Val537, Accession # NP_849155
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133308
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.