Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHA2/SLC9B2/NHEDC2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15957920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15957920 20 μL
NBP159579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15957920 Supplier Novus Biologicals Supplier No. NBP15957920UL

Rabbit Polyclonal Antibody

NHA2/SLC9B2/NHEDC2 Polyclonal specifically detects NHA2/SLC9B2/NHEDC2 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen NHA2/SLC9B2/NHEDC2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q86UD5
Gene Alias FLJ23984, Mitochondrial Na(+)/H(+) exchanger NHA2, mitochondrial sodium/hydrogen exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, Na+/H+ exchanger domain containing 2, NHA2NHE10, NHE domain-containing protein 2, Sodium/hydrogen exchanger-like domain-containing protein 2
Gene Symbols SLC9B2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to NHEDC2(Na+/H+ exchanger domain containing 2) The peptide sequence was selected from the C terminal of NHEDC2. Peptide sequence IFISFAWLPKATVQAAIGSVALDTARSHGEKQLEDYGMDVLTVAFLSILI.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133308
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.