Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHE3/SLC9A3, p Ser552 Antibody (14D5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013950 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013950 Supplier Novus Biologicals Supplier No. NBP280877
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NHE3/SLC9A3 (p Ser552) Monoclonal antibody specifically detects NHE3/SLC9A3 (p Ser552) in Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay, Knockdown.

Specifications

Antigen NHE3/SLC9A3 (p Ser552)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay, KnockDown
Classification Monoclonal
Clone 14D5
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-5 ug/ml, Simple Western : 1:1000, Flow Cytometry reported in scientific literature (PMID 21255452), ELISA : 1:100-1:2000, Immunohistochemistry : 1:50-1:1000, Immunocytochemistry/Immunofluorescence : 1:50-1:1000, Immunoprecipitation 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31276916), Knockdown Validated reported in scientific literature (PMID 31276916)
Formulation Tris-Glycine, 0.15 M NaCl
Gene Alias isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3
Gene Symbols SLC9A3
Host Species Mouse
Immunogen Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. [UniProt# P26433]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6550
Test Specificity Porcine reactivity reported in scientific literature (PMID: 29510506).
Target Species Human, Mouse, Rat, Pig, Opossum, Primate, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.