Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHE3/SLC9A3, p Ser605 Antibody (10A8), FITC, Novus Biologicals™
SDP

Catalog No. NB11074678F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11074678F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11074678F Supplier Novus Biologicals Supplier No. NB11074678F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NHE3/SLC9A3 (p Ser605)
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 10A8
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Gene Alias isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3
Gene Symbols SLC9A3
Host Species Mouse
Immunogen Synthetic peptide made to a region surrounding the Serine 605 portion of rat NHE3. [UniProt# P26433]
Molecular Weight of Antigen 85 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6550
Test Specificity This antibody detects NHE3 posphorylated at serine 605, and does not cross-react with the non-phosphopeptide.
Target Species Mouse, Rat, Pig, Opossum, Primate, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.