Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHE3/SLC9A3, p Ser605 Antibody (10A8), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339036 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339036 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339036 Supplier Novus Biologicals Supplier No. NB11074678MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NHE3/SLC9A3 Monoclonal antibody specifically detects NHE3/SLC9A3 in Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen NHE3/SLC9A3
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 10A8
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3
Host Species Mouse
Immunogen Synthetic peptide made to a region surrounding the Serine 605 portion of rat NHE3. [UniProt# P26433]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6550
Target Species Mouse, Rat, Pig, Opossum, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.