Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHE8 Antibody (7A11), HRP, Novus Biologicals™
SDP

Catalog No. NB11062091H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11062091H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11062091H Supplier Novus Biologicals Supplier No. NB11062091H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NHE8 Monoclonal specifically detects NHE8 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen NHE8
Applications Western Blot
Classification Monoclonal
Clone 7A11
Conjugate HRP
Dilution Western Blot
Gene Accession No. Q9Y2E8
Gene Alias DKFZp686C03237, KIAA0939DKFZp686C03237, MGC138418, Na(+)/H(+) exchanger 8, NHE-8, NHE8FLJ42500, sodium/hydrogen exchanger 8, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 8, solute carrier family 9 (sodium/hydrogen exchanger), member 8, Solute carrier family 9 member 8
Gene Symbols SLC9A8
Host Species Mouse
Immunogen A fusion protein containing the C-terminal 89 residues of human NHE8. [Swiss-Prot# Q9Y2E8]
Molecular Weight of Antigen 80 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23315
Test Specificity This is specific for the 85 kDa mature glycosylated form of the protein. It does not recognize the 55 kDa immature form.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.