Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHERF-1 Antibody (EBP-10), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB359205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359205 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359205 Supplier Novus Biologicals Supplier No. NB500488MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NHERF-1 Monoclonal antibody specifically detects NHERF-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen NHERF-1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EBP-10
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EBP50NHERF-1, ezrin-radixin-moesin binding phosphoprotein-50, Ezrin-radixin-moesin-binding phosphoprotein 50, Na(+)/H(+) exchange regulatory cofactor NHE-RF1, Na+/H+ exchange regulatory co-factor, NHERF1solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1, NHERFNPHLOP2, Regulatory cofactor of Na(+)/H(+) exchanger, sodium/hydrogen exchanger regulatory factor 1, Sodium-hydrogen exchanger regulatory factor 1, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 1, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1, Solute carrier family 9 isoform A3 regulatory factor 1
Host Species Mouse
Immunogen Bacterially produced recombinant full-length human NHERF1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9368
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.