Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NHERF-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300536 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300536 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300536 Supplier Novus Biologicals Supplier No. NB300536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

NHERF-1 Polyclonal specifically detects NHERF-1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen NHERF-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 10 μg/mL
Formulation PBS and 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. O14745
Gene Alias EBP50NHERF-1, ezrin-radixin-moesin binding phosphoprotein-50, Ezrin-radixin-moesin-binding phosphoprotein 50, Na(+)/H(+) exchange regulatory cofactor NHE-RF1, Na+/H+ exchange regulatory co-factor, NHERF1solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1, NHERFNPHLOP2, Regulatory cofactor of Na(+)/H(+) exchanger, sodium/hydrogen exchanger regulatory factor 1, Sodium-hydrogen exchanger regulatory factor 1, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 1, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1, Solute carrier family 9 isoform A3 regulatory factor 1
Gene Symbols SLC9A3R1
Host Species Rabbit
Immunogen Bacterially expressed, His-tagged human EBP50.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9368
Test Specificity Detects EBP 50 from human T84 and Calu3 cells as well as recombinant human protein.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.