Learn More
Description
Specifications
Specifications
| Antigen | NHLH1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | BHLHA35, bHLHa35HEN-1, helix-loop-helix protein 1, HEN1NSCL-1, nescient helix loop helix 1Class A basic helix-loop-helix protein 35, NSCL, NSCL1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human NHLH1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
