Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva NHS Mag Sepharose™

Catalog No. 45002159 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 μL
4 x 500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-159 500 μL
45-002-172 4 x 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-159 Supplier Cytiva Supplier No. 28944009
Add to Cart
Add to Cart

NHS Mag Sepharose magnetic beads for convenient enrichment of selected proteins using pull-down sample preparation techniques

NHS Mag Sepharose are magnetic beads designed for pull-down techniques enabling rapid capture and enrichment of selected proteins based on affinity. An antibody or other protein with desired affinity is covalently coupled to the medium via primary amines for the purpose of capture of target proteins of interest.

  • Visible and dense Sepharose based magnetic beads with N-hydroxysuccinimide (NHS) ligand, easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized capacity for enrichment/pull-down of target proteins - only low amounts of bound capture molecule
  • Flexible protocols with elution conditions optimized for downstream applications such as electrophoresis and MS

Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: N-hydroxysuccinimide
Ligand density: 8-14μmol/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% 2-propanol
Storage temperature: 4°C to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Bead Type NHS Mag Sepharose
Binding Capacity 8 to 14μM/mL Gel
Ligand Concentration 8 to 14μM/mL
Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100 μm
Quantity 500 μL
Content And Storage 4°C to 8°C
For Use With (Application) For pull-down techniques enabling rapid capture and enrichment of selected Proteins based on affinity
Ligand N-hydroxysuccinimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.