Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Niacinamide, USP, 98.5-101.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Niacinamide, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Ignition Residue | 0.001 |
| Loss on Drying | 0.005 |
| Melting Range | 128-131°C |
| Packaging | Poly Bottle |
| Quantity | 100 g |
| Percent Purity | 98.5 to 101.5% |
| Grade | USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction