Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niban Antibody, Novus Biologicals™
SDP

Catalog No. NB418161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB418161 25 μL
NBP188338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB418161 Supplier Novus Biologicals Supplier No. NBP18833825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Niban Polyclonal specifically detects Niban in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Niban
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias C1orf24, cell growth inhibiting protein 39, Cell growth-inhibiting gene 39 protein, chromosome 1 open reading frame 24, family with sequence similarity 129, member A, GIG39, NIBANFLJ38228, Protein FAM129A, protein Niban
Gene Symbols FAM129A
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ILLDETLKVIKEAAILKKHNLFEDNMALPSESVSSLTDLKPPTGSNQASPARRASAILPGVLGSETLSNEVFQESEEEKQPEVPSSLAKGESLSLPGP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116496
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.