Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NIBR 189

Click to view available options
Quantity:
10 mg
50 mg
Description
NIBR 189 is a potent and selective EBI2 (GPR183) receptor antagonist (IC50 values are 11 and 15 nM for human and mouse EBI2 receptors respectively). Exhibits selectivity for EBI2 over a panel of other receptors, enzymes and transporters. Blocks oxysterol-induced migration of U937 monocyte cells. Orally bioavailable.
Specifications
Specifications
Quantity | 10 mg |
Product Type | NIBR 189 |
Purity | >99% |
Molecular Weight (g/mol) | 429.31 |
Inhibitors | NIBR 189 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction