Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicalin Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP259116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259116 Supplier Novus Biologicals Supplier No. NBP259116

Rabbit Polyclonal Antibody

Nicalin Polyclonal specifically detects Nicalin in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Nicalin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation 20 ul/mg lysate
Formulation Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias NET59, nicalin, nicalin homolog (zebrafish), nicalin homolog (zebrefish), Nicastrin-like protein
Gene Symbols NCLN
Host Species Rabbit
Immunogen Maps to a region between 375 and 425
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56926
Target Species Human, Mouse
Content And Storage Store at 4C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.