Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicastrin Antibody (019), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB109615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109615 Supplier Novus Biologicals Supplier No. NBP289828UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Nicastrin Monoclonal specifically detects Nicastrin in Human samples. It is validated for Western Blot.

Specifications

Antigen Nicastrin
Applications Western Blot
Classification Monoclonal
Clone 19
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Nicastrin (NP_056146.1; Met1-Glu669).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23385
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.