Learn More
Nickel ammonium sulfate hexahydrate
Description
It is used in electroplating. Has limited use as dye mordants and is used in metal-finishing compositions. Nickel electrolyte for electroplating
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Physical Form | Large Crystals |
| Quantity | 500 g |
| Solubility Information | Soluble in ammonium sulphate. Insoluble in alcohol. Slightly soluble in water. |
| Formula Weight | 394.99 (286.90 Anhydrous) |
| Odor | Odorless |
| Appearance | Green crystals |
| Density | 1.923 |
| Chemical Name or Material | Nickel ammonium sulfate hexahydrate |
Safety and Handling
| Hazard Category | H302+H332-H317-H334-H341-H350i-H360D-H372 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 239-793-5 |
| RTECSNumber | WS6061000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.