Learn More
Nickel bis(trifluoromethylsulfonyl)imide
Description
Nickel(II) Bis(trifluoromethanesulfonyl)imide is used as Surface treatment, Synthesis and catalysis. It is also used as a ionic liquid in synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Green |
Physical Form | Solid |
Quantity | 1 g |
Linear Formula | [(CF3SO2)2N]2Ni |
UN Number | UN3261 |
Solubility Information | It is soluble in water. |
Formula Weight | 618.96 |
Sensitivity | Hygroscopic |
Chemical Name or Material | Nickel bis(trifluoromethylsulfonyl)imide |
Safety and Handling
Hazard Category | H314-H317-H335-H350 |
Hazard Statement | GHS H Statement H350-H314-H318-H317 May cause cancer. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.