Learn More
Nickel(II) carbonate, anhydrous, 98%
Description
Nickel(II) carbonate is used in sulphamate baths, metal phosphating, electroplating and ceramic applications. It acts as a precursor to catalysts and an intermediate in the hydrometallurgical purification of nickel from its ores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Beige |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 98% |
| Solubility Information | Soluble in dilute acids. Insoluble in cold water. |
| Formula Weight | 118.72 |
| Appearance | Light powder |
| Chemical Name or Material | Nickel(II) carbonate, anhydrous |
Safety and Handling
| Hazard Category | H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372 |
| Hazard Statement | Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c |
| DOTInformation | Hazard Class: 9, Packing Group: III |
| EINECSNumber | 222-068-2 |
| RTECSNumber | QR6200000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.