Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel (II) Chloride Hexahydrate, ACS Reagent, Reagents

Catalog No. p-200021127
Click to view available options
Quantity:
125 g

Catalog No. 01-337-882

Supplier: Reagents Holdings Llc C1255500125D2

Only null left
Add to Cart
Add to Cart

Nickel (II) Chloride Hexahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2H12NiO6
Molecular Weight (g/mol) 237.68
InChI Key LAIZPRYFQUWUBN-UHFFFAOYSA-L
Synonym Nickel(II) Chloride Hexahydrate, Nickelous Chloride Hexahydrate
IUPAC Name nickel(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ni++]

Specifications

Boiling Point 973°C
Color Green
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 3.55
Chemical Name or Material Nickel (II) Chloride Hexahydrate, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.