Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) hydroxide, Ni 61%

Catalog No. p-7049020
Click to view available options
Quantity:
250 g
1 kg

Catalog No. AA1251730

Supplier: Thermo Scientific Chemicals 01251730

Only null left
Add to Cart

CAS: 12054-48-7 | H2NiO2 | 92.71 g/mol

Due to its redox behavior, it finds use in rechargeable battery electrodes. It is useful for the storage of electrochemical energy, with potential applications in car batteries. It is a potential material for several biological applications, for example in chromosomal DNA quantification assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12054-48-7
Molecular Formula H2NiO2
Molecular Weight (g/mol) 92.71
MDL Number MFCD00011140
InChI Key BFDHFSHZJLFAMC-UHFFFAOYSA-L
IUPAC Name nickel(2+) dihydroxide
SMILES [OH-].[OH-].[Ni++]

Specifications

Melting Point 230°C (decomposition)
Color Green
Physical Form Powder
Quantity 250 g
Linear Formula Ni(OH)2
UN Number UN3077
Solubility Information Insoluble in water. Soluble in dilute acid.
Formula Weight 92.7
Concentration or Composition (by Analyte or Components) Ni 61%
Odor Odorless
Density 4.1 g/mL
Chemical Name or Material Nickel(II) hydroxide
Show More Show Less
Hazard Category H302+H332-H315-H317-H319-H334-H340-H350i-H360-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 235-008-5
RTECSNumber QR7040000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.