Learn More
Nickel(II) sulfate hexahydrate, 99.97% min (metals basis)
Description
Nickel(II) sulfate hexahydrate is mainly used in Ni plating, as mordant in dyeing and in printing textiles, as a blackening agent for zinc and brass and in the fabrication of organic nickel salts. It is used as an effective catalyst for gas-phase dehydration of glycerol to acrolein. Nickel sulfate hexahydrate crystal is an efficient ultraviolet filter material. Columns used in biochemistry and molecular biology can be regenerated with nickel sulfate. It is used to prepare nickel carbonate, a precursor to pigments and nickel-based catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Physical Form | Lump |
| Quantity | 50 g |
| Assay Percent Range | 99.97% min. (metals basis) |
| Solubility Information | Very soluble in water,ethanol and ammonia. |
| Formula Weight | 262.86 (154.77 Anhydrous) |
| Odor | Odorless |
| Density | 2.07 g/cm3 at 20°C |
| Chemical Name or Material | Nickel(II) sulfate hexahydrate |
Safety and Handling
| Hazard Category | H302-H315-H317-H319-H332-H334-H335-H341-H350i-H360D-H372 |
| Hazard Statement | Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c |
| DOTInformation | DOT Class: 9; Packaging Group: III |
| EINECSNumber | 232-104-9 |
| RTECSNumber | QR9600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.