Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ β-Nicotinamide Adenine Dinucleotide, Oxidized Form, MP Biomedicals™

Catalog No. MP021947145 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021947145 500 mg
MP021947141 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021947145 Supplier MP Biomedicals™ Supplier No. 02194714.5
Add to Cart
Add to Cart

This is an ultrapure NAD, chromatographically purified to remove trace inhibitors.

β-NAD, a pyridine nucleotide and biologically active form of nicotinic acid, is a coenzyme necessary for the catalytic reaction of certain enzymes. It occurs in living cells primarily in the oxidized state. It serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols. NAD usually acts as a hydrogen acceptor, forming NADH which then serves as a hydrogen donor in the respiratory chain.

  • Electron acceptor.
  • β-NAD is a carrier for the hydride ion, forming b-NADH.
  • Hydride ion is enzymatically removed from a substrate molecule by the action of dehydrogenases such as malic dehydrogenase and lactic dehydrogenase.
  • Such enzymes catalyze the reversible transfer of a hydride ion from malate or lactate to b-NAD to form the reduced product, b-NADH.
  • Unlike b-NAD which has no absorbance at 340nm, b-NADH absorbs at 340nm (EmM = 6.22).
  • The increase in absorbance at 340nm with the formation of b-NADH is the basis for measurement of activity of many enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Grade Cell Culture
Purity ≥98%
pH 2 to 4 (1% aq soln)
Content And Storage 0°C, Desiccate
Quantity 500 mg
Molecular Weight (g/mol) 663.4 (anhyd.)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.