Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ β-Nicotinamide Adenine Dinucleotide, Oxidized Form, MP Biomedicals™

Catalog No. p-7083035
Click to view available options
Quantity:
100 mg
500 mg

This is an ultrapure NAD, chromatographically purified to remove trace inhibitors.

β-NAD, a pyridine nucleotide and biologically active form of nicotinic acid, is a coenzyme necessary for the catalytic reaction of certain enzymes. It occurs in living cells primarily in the oxidized state. It serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols. NAD usually acts as a hydrogen acceptor, forming NADH which then serves as a hydrogen donor in the respiratory chain.

  • Electron acceptor.
  • β-NAD is a carrier for the hydride ion, forming b-NADH.
  • Hydride ion is enzymatically removed from a substrate molecule by the action of dehydrogenases such as malic dehydrogenase and lactic dehydrogenase.
  • Such enzymes catalyze the reversible transfer of a hydride ion from malate or lactate to b-NAD to form the reduced product, b-NADH.
  • Unlike b-NAD which has no absorbance at 340nm, b-NADH absorbs at 340nm (EmM = 6.22).
  • The increase in absorbance at 340nm with the formation of b-NADH is the basis for measurement of activity of many enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Grade Cell Culture
Purity ≥98%
pH 2 to 4 (1% aq soln)
Content And Storage 0°C, Desiccate
Quantity 100 mg
Molecular Weight (g/mol) 663.4 (anhyd.)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.