Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinamide N-Methyltransferase/NNMT Antibody (OTI3D8), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP272984UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272984UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272984UV Supplier Novus Biologicals Supplier No. NBP272984UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen Nicotinamide N-Methyltransferase/NNMT
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Monoclonal
Clone OTI3D8
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.1.1.1, nicotinamide N-methyltransferase
Gene Symbols NNMT
Host Species Mouse
Immunogen Full length recombinant protein of human Nicotinamide N-Methyltransferase/NNMT (NP_006160) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4837.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.