Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acetylcholine R alpha 1/CHRNA1 Antibody (mAb 192), PerCP, Novus Biologicals™
SDP

Catalog No. NB342888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342888 Supplier Novus Biologicals Supplier No. NBP281070PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Nicotinic Acetylcholine R alpha 1/CHRNA1 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 1/CHRNA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Neutralization

Specifications

Antigen Nicotinic Acetylcholine R alpha 1/CHRNA1
Applications Western Blot, ELISA, Immunocytochemistry, Neutralization
Classification Monoclonal
Clone mAb 192
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Block/Neutralize
Formulation PBS
Gene Alias acetylcholine receptor subunit alpha, ACHRA, ACHRD, CHNRA, cholinergic receptor, nicotinic, alpha 1 (muscle), CHRNA, CMS2A, FCCMS, nicotinic acetylcholine receptor alpha subunit, nicotinic cholinergic receptor alpha 1, nicotinic, alpha polypeptide 1 (muscle), SCCMS
Host Species Rat
Immunogen Human muscle AChR.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1134
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.